This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ERLEC1 SIRNA Assays ›
Gene Symbol:
Erlec1
Gene Name:
endoplasmic reticulum lectin 1
Gene Aliases:
4933407N01Rik
Chromosome Location:
Chr.11: 30865632 - 30954173 on Build GRCm38
UniGene ID:
Mm.294641
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227704
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Erlec1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025745.3 NP_080021.3 7 680
GenBank mRNA AK002471.1 - 7 671
AK077120.1 - 7 673
AK133874.1 - 7 681
BC018468.1 - 7 690 3591901

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106023, XM_006251615, FQ228963, FQ232613.

Gene Ontology Categories:

Function(s) Process(es)

ER-associated ubiquitin-dependent protein catabolic process
negative regulation of retrograde protein transport, ER to cytosol
glycoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0