This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM186 SIRNA Assays ›
Gene Symbol:
Tmem186
Gene Name:
transmembrane protein 186
Gene Aliases:
2810440M13Rik, 4432406C05Rik, AI648175, AW210586
Chromosome Location:
Chr.16: 8633731 - 8637701 on Build GRCm38
UniGene ID:
Mm.10068
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227647
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem186
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025708.4 NP_079984.2 2 816
GenBank mRNA AK009236.1 - 2 653
AK013147.1 - 2 645
AK014478.1 - 2 817
AK033686.1 - 2 654
AK088656.1 - 2 826
AK159727.1 - 2 802
AK159782.1 - 2 802
BC027261.1 - 2 646 2649343
BC058268.1 - 1 1878

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header