This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZMAT2 SIRNA Assays ›
Gene Symbol:
Zmat2
Gene Name:
zinc finger, matrin type 2
Gene Aliases:
2610510D14Rik, 2900082I05Rik
Chromosome Location:
Chr.18: 36793923 - 36799660 on Build GRCm38
UniGene ID:
Mm.251155
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227476
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zmat2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025594.3 NP_079870.1 4 314
GenBank mRNA AK008717.1 - 4 307
AK012091.1 - 4 310
BC024881.1 - 4 314 4022326
BC089501.1 - 4 309 6594761

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleic acid binding
DNA binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0