This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALG5 SIRNA Assays ›
Gene Symbol:
Alg5
Gene Name:
asparagine-linked glycosylation 5 (dolichyl-phosphate beta-glucosyltransferase)
Gene Aliases:
1500026A19Rik, 2600005J22Rik, AV009521, AV170001, AW545564
Chromosome Location:
Chr.3: 54735539 - 54749795 on Build GRCm38
UniGene ID:
Mm.144218
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227335
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Alg5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025442.3 NP_079718.1 8 693
GenBank mRNA AK003818.1 - 8 674
AK005296.1 - 8 694
AK006206.1 - 8 662
AK161187.1 - 672
BC027160.1 - 8 653 4946430

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

determination of left/right symmetry
dolichyl-phosphate beta-glucosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0