This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SF3B6 SIRNA Assays ›
Gene Symbol:
Sf3b6
Gene Name:
splicing factor 3B, subunit 6
Gene Aliases:
0610009D07Rik, 6030419K15Rik, AV001342, Sf3b14
Chromosome Location:
Chr.12: 4817608 - 4827659 on Build GRCm38
UniGene ID:
Mm.102627
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227230
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sf3b6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025323.2 NP_079599.1 3 343
GenBank mRNA AK002375.1 - 3 391
AK003683.1 - 3 399
AK151648.1 - 3 386
BC019535.1 - 3 343 4458983

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

blastocyst formation
mRNA processing
RNA splicing
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0