This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TSC1 SIRNA Assays ›
Gene Symbol:
Tsc1
Gene Name:
tuberous sclerosis 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 28641190 - 28691172 on Build GRCm38
UniGene ID:
Mm.224354
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS227145
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tsc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289575.1 NP_001276504.1 22 3104
NM_001289576.1 NP_001276505.1 22 3086
NM_022887.4 NP_075025.2 22 3072
GenBank mRNA AB047561.1 - 20 2871
AJ271912.1 - 22 3096
AK030318.1 - 22 3069
AK047656.1 - 1 55
AK122229.1 - 8 1396
AK147428.1 - 22 3077
BC052399.1 - 21 3025 5720967

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_021854, XM_006233844, XM_006233845, XM_006233849, XM_006233847, and others.

Gene Ontology Categories:

Function(s) Process(es)

kidney development
neural tube closure
regulation of cell-matrix adhesion
rRNA export from nucleus
regulation of translation
potassium ion transport
cell-matrix adhesion
nervous system development
negative regulation of cell proliferation
embryo development
negative regulation of translation
hippocampus development
cerebral cortex development
cell projection organization
negative regulation of TOR signaling
response to insulin
regulation of actin cytoskeleton organization
myelination
regulation of GTPase activity
regulation of phosphoprotein phosphatase activity
negative regulation of cell size
regulation of protein kinase activity
glucose import
negative regulation of insulin receptor signaling pathway
synapse organization
protein stabilization
protein heterooligomerization
regulation of stress fiber assembly
regulation of cell cycle
regulation of focal adhesion assembly
positive regulation of focal adhesion assembly
cardiac muscle cell differentiation
activation of GTPase activity
protein binding
GTPase regulator activity
GTPase activating protein binding
protein N-terminus binding
chaperone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0