This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CACNA2D2 SIRNA Assays ›
Gene Symbol:
Cacna2d2
Gene Name:
calcium channel, voltage-dependent, alpha 2/delta subunit 2
Gene Aliases:
Cacna2d, a2d2, du, mKIAA0558, td, torpid
Chromosome Location:
Chr.9: 107399880 - 107529346 on Build GRCm38
UniGene ID:
Mm.273084
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS226426
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cacna2d2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174047.1 NP_001167518.1 39 3575
NM_001174048.1 NP_001167519.1 38 3557
NM_001174049.1 NP_001167520.1 38 3548
NM_001174050.1 NP_001167521.1 38 3551
NM_020263.3 NP_064659.2 39 3569
GenBank mRNA AB093246.1 - 38 3212
AF169633.1 - 7 587
AF247139.1 - 40 3829
AF247141.1 - 38 3170
AK044603.1 - 38 3367
AK161839.1 - 12 1028
AK164143.1 - 17 1395
AY502107.1 - 38 3476
BC056389.1 - 39 3569 6854935
BC158058.1 - 38 3379 9007358

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
calcium ion transport
neuromuscular junction development
regulation of ion transmembrane transport
regulation of multicellular organism growth
positive regulation of organ growth
muscle fiber development
regulation of calcium ion transport
rhythmic synaptic transmission
calcium ion transmembrane transport
voltage-gated ion channel activity
voltage-gated calcium channel activity
calcium channel regulator activity
calcium channel activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0