This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRPC4AP SIRNA Assays ›
Gene Symbol:
Trpc4ap
Gene Name:
transient receptor potential cation channel, subfamily C, member 4 associated protein
Gene Aliases:
4833429F06Rik, D2Ertd113e, Trrp4ap, mFLJ00177, truss
Chromosome Location:
Chr.2: 155634277 - 155692384 on Build GRCm38
UniGene ID:
Mm.234944
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS226128
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trpc4ap
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163452.1 NP_001156924.1 15, 16 1902
NM_019828.2 NP_062802.2 15, 16 1878
GenBank mRNA AF093589.1 - 11, 12 1374
AF130458.1 - 15, 16 1826
AF509881.1 - 15, 16 1824
AK156047.1 - 15, 16 1851
AK170085.1 - 15, 16 1840
AK171404.1 - 15, 16 1851
AK190008.1 - 1, 2 46
AK191521.1 - 1, 2 46
AK192276.1 - 1, 2 46
AK196821.1 - 1, 2 46
AK197729.1 - 1, 2 46
AK197922.1 - 1, 2 46
AK198111.1 - 1, 2 46
AK200382.1 - 1, 2 46
AK200874.1 - 1, 2 46
AK201803.1 - 1, 2 46
AK202669.1 - 1, 2 46
AK203016.1 - 1, 2 46
AK220239.1 - 15, 16 1878
BC003931.1 - 8, 9 838
BC033274.1 - 15, 16 1830 3673725
BC057330.1 - 15, 16 1860 6816155

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001100748, AY724540, BC166758, BC087662, BC105861.

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0