This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CACNA1F SIRNA Assays ›
Gene Symbol:
Cacna1f
Gene Name:
calcium channel, voltage-dependent, alpha 1F subunit
Gene Aliases:
A930034B14, Cav1.4, Sfc17, nerg1, nob2
Chromosome Location:
Chr.X: 7606982 - 7635196 on Build GRCm38
UniGene ID:
Mm.289647
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS225744
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cacna1f
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019582.2 NP_062528.2 21, 22 2746
GenBank mRNA AF192497.1 - 21, 22 2757
AJ579852.2 - 21, 22 2703
BC043482.1 - 18, 19 3295
BC051413.1 - 21, 22 2720

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
calcium ion transport
cellular calcium ion homeostasis
axonogenesis
visual perception
regulation of ion transmembrane transport
T cell homeostasis
dendrite morphogenesis
regulation of T cell receptor signaling pathway
response to stimulus
detection of light stimulus involved in visual perception
transmembrane transport
retina development in camera-type eye
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
ion channel activity
voltage-gated ion channel activity
voltage-gated calcium channel activity
calcium channel activity
high voltage-gated calcium channel activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0