This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SVS2 SIRNA Assays ›
Gene Symbol:
Svs2
Gene Name:
seminal vesicle secretory protein 2
Gene Aliases:
BB115391, Semg1, SvsII
Chromosome Location:
Chr.2: 164235929 - 164238341 on Build GRCm38
UniGene ID:
Mm.99395
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS225442
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Svs2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017390.4 NP_059086.3 2 772
GenBank mRNA AK020661.1 - 2 216
AK035204.1 - 2 697
AK040724.1 - 2 698
AK090151.1 - 2 694
AK090302.1 - 2 698
AK136679.1 - 2 696
AK144285.1 - 2 697
AK144389.1 - 2 697
AK144429.1 - 2 697
AK144524.1 - 2 694
AK165704.1 - 2 697
AK165713.1 - 2 697
AK165727.1 - 2 694
AK165753.1 - 2 697
AK165825.1 - 2 697
BC107275.1 - 2 697 40052650
BC107276.1 - 2 697

Back To Top

More Information


Gene Ontology Categories:

Function(s)

fertilization
sperm capacitation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header