This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC46A1 SIRNA Assays ›
Gene Symbol:
Slc46a1
Gene Name:
solute carrier family 46, member 1
Gene Aliases:
1110002C08Rik, D11Ertd18e, HCP1, Pcft
Chromosome Location:
Chr.11: 78465701 - 78471945 on Build GRCm38
UniGene ID:
Mm.131618
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS225195
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc46a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026740.2 NP_081016.2 2 906
GenBank mRNA AK003278.1 - 2 889
AK170505.1 - 2 899
AK220201.1 - 2 866
BC024522.1 - 1 42
BC057976.1 - 2 891 5052851

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
folic acid transport
heme transport
methotrexate transport
transmembrane transport
transporter activity
folic acid binding
folic acid transporter activity
heme transporter activity
methotrexate transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0