This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HS6ST1 SIRNA Assays ›
Gene Symbol:
Hs6st1
Gene Name:
heparan sulfate 6-O-sulfotransferase 1
Gene Aliases:
6Ost1
Chromosome Location:
Chr.1: 36068400 - 36106446 on Build GRCm38
UniGene ID:
Mm.213566
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS224976
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hs6st1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015818.2 NP_056633.2 2 965
GenBank mRNA AB024566.1 - 2 956
AK144809.1 - 3 885
AK157556.1 - 2 829
AK165161.1 - 1 2005
AK165385.1 - 2 368
AK166962.1 - 70
AK167151.1 - 2 394
BC052316.1 - 2 965 6391204

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, enzymatic modification
lung alveolus development
neuron development
labyrinthine layer blood vessel development
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0