This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RXFP1 SIRNA Assays ›
Gene Symbol:
Rxfp1
Gene Name:
relaxin/insulin-like family peptide receptor 1
Gene Aliases:
Gm1018, Lgr7
Chromosome Location:
Chr.3: 79641600 - 79737951 on Build GRCm38
UniGene ID:
Mm.331086
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS221829
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rxfp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212452.1 NP_997617.1 14 1101
GenBank mRNA AK054129.1 - 649
AK137655.1 - 13 1082
AY509975.1 - 14 1101

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
parturition
cell differentiation
extracellular matrix organization
lung connective tissue development
nipple morphogenesis
signal transducer activity
G-protein coupled receptor activity
hormone binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header