This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZSCAN10 SIRNA Assays ›
Gene Symbol:
Zscan10
Gene Name:
zinc finger and SCAN domain containing 10
Gene Aliases:
BC068284, Zfp206, Zkscan10, Znf206
Chromosome Location:
Chr.17: 23599751 - 23611019 on Build GRCm38
UniGene ID:
Mm.270315
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS221191
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zscan10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033425.4 NP_001028597.2 6 1003
NM_001289481.1 NP_001276410.1 6 1003
NM_001289482.1 NP_001276411.1 4 673
NM_001289483.1 NP_001276412.1 6 499
NM_001289484.1 NP_001276413.1 6 868
GenBank mRNA AK141259.1 - 6 974
BC068284.1 - 5 676
BC145638.1 - 5 836 9054214
CD539380.1 - 2 307
DQ323929.1 - 6 971
DQ323930.1 - 6 971
DQ323931.1 - 4 641
EF152498.1 - 5 818
EF152499.1 - 6 463
EF152501.1 - 4 595
EF152502.1 - 4 595
EF152503.1 - 2 472
EF152504.1 - 3 529

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription, DNA-templated
stem cell differentiation
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0