This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPIP5K1 SIRNA Assays ›
Gene Symbol:
Ppip5k1
Gene Name:
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases:
B430315C20Rik, Hisppd2a, mKIAA0377
Chromosome Location:
Chr.2: 121310557 - 121355636 on Build GRCm38
UniGene ID:
Mm.386076
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS220662
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppip5k1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178795.4 NP_848910.3 28 3438
GenBank mRNA AF502585.1 - 28 3440
AK122264.1 - 28 3404
BC065138.1 - 15 1670
BC138351.1 - 28 3486 8861371

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001080783, XM_006234840, XM_006234841, XM_008762170, XM_006234844, and others.

Gene Ontology Categories:

Function(s) Process(es)

inositol metabolic process
phosphorylation
nucleotide binding
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
kinase activity
transferase activity
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0