This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OCRL SIRNA Assays ›
Gene Symbol:
Ocrl
Gene Name:
oculocerebrorenal syndrome of Lowe
Gene Aliases:
9530014D17Rik, BB143339, OCRL1
Chromosome Location:
Chr.X: 47912456 - 47966428 on Build GRCm38
UniGene ID:
Mm.210343
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS220503
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ocrl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177215.3 NP_796189.2 18 2135
GenBank mRNA AK035315.1 - 18 2138
AK041963.1 - 18 2137
AK044320.1 - 16 1891
AK047906.1 - 18 2154
BC043709.1 - 5 517
BC068146.1 - 18 2135 30533468

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
signal transduction
cell projection organization
cilium assembly
regulation of GTPase activity
positive regulation of GTPase activity
phosphatidylinositol dephosphorylation
phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
GTPase activator activity
hydrolase activity
Rac GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0