This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PHF8 SIRNA Assays ›
Gene Symbol:
Phf8
Gene Name:
PHD finger protein 8
Gene Aliases:
9830141C09Rik, mKIAA1111
Chromosome Location:
Chr.X: 151520612 - 151633859 on Build GRCm38
UniGene ID:
Mm.17156
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS220485
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phf8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113354.1 NP_001106825.1 7 1625
NM_177201.5 NP_796175.1 7 1625
GenBank mRNA AK036609.1 - 7 999
AK040943.1 - 7 994
AK040969.1 - 7 1626
AK122447.1 - 6 716
AK134137.1 - 7 962
AK147939.1 - 7 1012
BC138899.1 - 7 894 8861921

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108253, XM_008773119, XM_006256792, XM_006256796, XM_006256794, and others.

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
histone H3-K9 demethylation
histone H4-K20 demethylation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase I promoter
oxidation-reduction process
negative regulation of chromatin silencing at rDNA
histone H3-K36 demethylation
histone H3-K27 demethylation
chromatin binding
iron ion binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
histone demethylase activity
histone demethylase activity (H3-K9 specific)
methylated histone binding
histone demethylase activity (H4-K20 specific)
histone demethylase activity (H3-K36 specific)
histone demethylase activity (H3-K27 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0