This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AGMO SIRNA Assays ›
Gene Symbol:
Agmo
Gene Name:
alkylglycerol monooxygenase
Gene Aliases:
A530016O06Rik, AI790538, Tmem195
Chromosome Location:
Chr.12: 37241629 - 37581932 on Build GRCm38
UniGene ID:
Mm.485540
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS220176
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Agmo
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178767.5 NP_848882.2 3 685
GenBank mRNA AK039191.1 - 3 256
AK040702.1 - 3 686
AK050246.1 - 3 546
AK163155.1 - 3 466

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid biosynthetic process
membrane lipid metabolic process
ether lipid metabolic process
oxidation-reduction process
iron ion binding
oxidoreductase activity
glyceryl-ether monooxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header