This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GYG SIRNA Assays ›
Gene Symbol:
Gyg
Gene Name:
glycogenin
Gene Aliases:
AU017667, Gyg1
Chromosome Location:
Chr.3: 20122084 - 20155266 on Build GRCm38
UniGene ID:
Mm.6375
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS219521
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gyg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013755.3 NP_038783.1 2 211
GenBank mRNA AA553085.1 - 2 210
AF114031.1 - 2 146
AK159576.1 - 2 192
AK201125.1 - 1 119
AK205608.1 - 2 109
AK209105.1 - 2 128
AK209409.1 - 2 109
BC029903.1 - 2 172 4487769

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glycogen biosynthetic process
glucose binding
glycogenin glucosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0