This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAPK1 SIRNA Assays ›
Gene Symbol:
Mapk1
Gene Name:
mitogen-activated protein kinase 1
Gene Aliases:
9030612K14Rik, AA407128, AU018647, C78273, ERK, Erk2, MAPK2, PRKM2, Prkm1, p41mapk, p42mapk
Chromosome Location:
Chr.16: 16983382 - 17047453 on Build GRCm38
UniGene ID:
Mm.196581
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS218549
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mapk1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038663.1 NP_001033752.1 7 1176
NM_011949.3 NP_036079.1 7 1176
GenBank mRNA AK035386.1 - 7 1150
AK048127.1 - 7 1178
AK087925.1 - 7 1151
AK132241.1 - 7 1164
AK148853.1 - 7 1138
AK211534.1 - 2 135
BC006708.1 - 2 222
BC058258.1 - 7 1119 6468233
D10939.1 - 7 991
X58712.1 - 7 949

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
MAPK import into nucleus
transcription, DNA-templated
protein phosphorylation
apoptotic process
cellular response to DNA damage stimulus
cell cycle
signal transduction
response to toxic substance
organ morphogenesis
positive regulation of peptidyl-threonine phosphorylation
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
sensory perception of pain
cytosine metabolic process
lipopolysaccharide-mediated signaling pathway
response to lipopolysaccharide
regulation of stress-activated MAPK cascade
mammary gland epithelial cell proliferation
intracellular signal transduction
ERBB signaling pathway
response to exogenous dsRNA
response to estrogen
negative regulation of cell differentiation
positive regulation of translation
T cell receptor signaling pathway
B cell receptor signaling pathway
regulation of sequence-specific DNA binding transcription factor activity
regulation of cytoskeleton organization
long-term synaptic potentiation
lung morphogenesis
trachea formation
labyrinthine layer blood vessel development
ERK1 and ERK2 cascade
response to epidermal growth factor
cellular response to organic substance
caveolin-mediated endocytosis
regulation of Golgi inheritance
cellular response to granulocyte macrophage colony-stimulating factor stimulus
regulation of early endosome to late endosome transport
nucleotide binding
phosphotyrosine binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
RNA polymerase II carboxy-terminal domain kinase activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0