This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAP3K12 SIRNA Assays ›
Gene Symbol:
Map3k12
Gene Name:
mitogen-activated protein kinase kinase kinase 12
Gene Aliases:
DLK, MUK, Zpk
Chromosome Location:
Chr.15: 102497647 - 102518053 on Build GRCm38
UniGene ID:
Mm.172897
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS218523
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Map3k12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163643.1 NP_001157115.1 16 3024
NM_009582.4 NP_033608.3 14 2848
GenBank mRNA AK029882.1 - 14 2657
AK034374.1 - 16 2840
AK081623.1 - 9 1516
AK166435.1 - 16 2944
BC047158.1 - 13 2642 6494216
BC057572.1 - 16 2848 6401321
U14636.1 - 14 2680
U23789.1 - 13 2638

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006301, NM_001193511, AK314530, AB209453, AK094195, and others.
This siRNA also targets the following transcripts in Rat: NM_013055, XM_008765770, XM_006242419, XM_006242421, XM_006242424, and others.

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
protein phosphorylation
JNK cascade
phosphorylation
histone phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
protein autophosphorylation
mitophagy in response to mitochondrial depolarization
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0