siRNA Details
Gene Symbol: Olfr1089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011771.2 | NP_001011771.2 | 1 | 737 |

Gene Symbol: Olfr1089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011771.2 | NP_001011771.2 | 1 | 737 |