This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HPS5 SIRNA Assays ›
Gene Symbol:
Hps5
Gene Name:
Hermansky-Pudlak syndrome 5 homolog (human)
Gene Aliases:
AI646796, AL022647, C85120, haze, hz, maroon, mr, ru-2, ru2
Chromosome Location:
Chr.7: 46760466 - 46796064 on Build GRCm38
UniGene ID:
Mm.379442
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS217366
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hps5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005247.2 NP_001005247.2 22 3759
NM_001005248.2 NP_001005248.2 22 3735
NM_001167864.1 NP_001161336.1 22 3660
GenBank mRNA AF534396.1 - 22 3556
AF534397.1 - 22 3532
AF534398.1 - 20 3121
AF534399.1 - 23 3687
AK129265.1 - 22 3682
AK140780.1 - 11 1941
AK141088.1 - 21 4384
AK154612.1 - 22 3489
AK157182.1 - 22 3525
AK161039.1 - 13 2130
BC046405.1 - 17 2738
BC046443.1 - 22 3373
BC082542.1 - 22 3660 6405811

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

organelle organization
blood coagulation
pigmentation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0