This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLHL21 SIRNA Assays ›
Gene Symbol:
Klhl21
Gene Name:
kelch-like 21
Gene Aliases:
1810045K06Rik, D330008A20, mKIAA0469
Chromosome Location:
Chr.4: 152008891 - 152017677 on Build GRCm38
UniGene ID:
Mm.22306
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS216667
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klhl21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033352.3 NP_001028524.1 2 1361
GenBank mRNA AK052203.1 - 2 1336
AK154493.1 - 2 1362
AK198752.1 - 1 295
BC060201.1 - 2 1350
BC141158.1 - 2 1335 9055770
BC141159.1 - 2 1335 9055771

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107996.

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
protein ubiquitination
regulation of cytokinesis
chromosome passenger complex localization to spindle midzone
cell division
ubiquitin-protein transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0