This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM4922 SIRNA Assays ›
Gene Symbol:
Gm4922
Gene Name:
predicted gene 4922
Gene Aliases:
4933423E17, EG237300
Chromosome Location:
Chr.10: 18777168 - 18786793 on Build GRCm38
UniGene ID:
Mm.444821
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS215315
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm4922
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177706.4 NP_808374.1 3 1367
GenBank mRNA AK030207.1 - 3 1369
BC125502.1 - 2 1087 40130017
BC125506.1 - 2 1087 40130021

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
intracellular signal transduction
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header