This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC17A5 SIRNA Assays ›
Gene Symbol:
Slc17a5
Gene Name:
solute carrier family 17 (anion/sugar transporter), member 5
Gene Aliases:
4631416G20Rik, 4732491M05, AST, ISSD, NSD, SD, SIALIN, SIASD, SLD
Chromosome Location:
Chr.9: 78536487 - 78588065 on Build GRCm38
UniGene ID:
Mm.46932
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS215084
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc17a5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276452.1 NP_001263381.1 7 946
NM_172773.3 NP_766361.1 7 1024
GenBank mRNA AK029102.1 - 7 1007
AK169868.1 - 7 973
BC058785.1 - 7 916 6309785

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
amino acid transport
sialic acid transport
transmembrane transport
anion transmembrane transport
organic acid transmembrane transport
sialic acid transmembrane transporter activity
symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header