This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM161A SIRNA Assays ›
Gene Symbol:
Tmem161a
Gene Name:
transmembrane protein 161A
Gene Aliases:
AI428876, BB161850, BC021367
Chromosome Location:
Chr.8: 70172329 - 70183681 on Build GRCm38
UniGene ID:
Mm.23488
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS214837
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem161a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293797.1 NP_001280726.1 8 822
NM_145597.4 NP_663572.1 9 926
GenBank mRNA AK032711.1 - 5 442
AK049979.1 - 9 925
AK082916.1 - 8 823
AK088569.1 - 8 812
AK147962.1 - 9 919
BC021367.1 - 9 926 5030434

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to retinoic acid
cellular response to oxidative stress
cellular response to UV
positive regulation of DNA repair
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0