This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP574 SIRNA Assays ›
Gene Symbol:
Zfp574
Gene Name:
zinc finger protein 574
Gene Aliases:
A630056B21Rik, Znf574
Chromosome Location:
Chr.7: 25075684 - 25083492 on Build GRCm38
UniGene ID:
Mm.41568
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS214559
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp574
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168506.1 NP_001161978.1 2 222
NM_175477.4 NP_780686.1 2 277
GenBank mRNA AK042074.1 - 2 240
AK053744.1 - 2 163
AK143997.1 - 2 223
AK170243.1 - 2 128
BC021833.2 - 2 222
BC054064.1 - 2 178
BC059044.1 - 2 148 6816316
BC094354.1 - 2 201
BY087218.1 - 2 278

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleic acid binding
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0