This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC5A9 SIRNA Assays ›
Gene Symbol:
Slc5a9
Gene Name:
solute carrier family 5 (sodium/glucose cotransporter), member 9
Gene Aliases:
AI159731, SGLT4
Chromosome Location:
Chr.4: 111875374 - 111902796 on Build GRCm38
UniGene ID:
Mm.26630
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS214017
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc5a9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145551.4 NP_663526.3 5 550
GenBank mRNA AK029158.1 - 5 551
AK033425.1 - 4 480
AK050696.1 - 4 430

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108974, XM_006238598.

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
transmembrane transport
transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0