This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YIPF1 SIRNA Assays ›
Gene Symbol:
Yipf1
Gene Name:
Yip1 domain family, member 1
Gene Aliases:
C030002N13Rik
Chromosome Location:
Chr.4: 107314254 - 107359823 on Build GRCm38
UniGene ID:
Mm.29118
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS214002
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Yipf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205156.1 NP_001192085.1 5 475
NM_145550.3 NP_663525.1 6 556
GenBank mRNA AK081205.1 - 6 543
AK157536.1 - 6 543
AK212572.1 - 1 22
AK214559.1 - 1 25
BC009080.1 - 6 488 2655774

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
mitophagy in response to mitochondrial depolarization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0