This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YTHDF1 SIRNA Assays ›
Gene Symbol:
Ythdf1
Gene Name:
YTH domain family 1
Gene Aliases:
2210410K23Rik, 8030473O16
Chromosome Location:
Chr.2: 180904377 - 180920936 on Build GRCm38
UniGene ID:
Mm.30205
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS213680
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ythdf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173761.3 NP_776122.1 4 571
GenBank mRNA AK033241.1 - 5 661
AK046768.1 - 4 585
AK075846.1 - 4 550
AK172582.1 - 4 545
BC061479.1 - 4 542 6334535
BC065050.1 - 4 571 6418891

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024756, BC097360.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
RNA binding
poly(A) RNA binding
N6-methyladenosine-containing RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0