This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRP4 SIRNA Assays ›
Gene Symbol:
Lrp4
Gene Name:
low density lipoprotein receptor-related protein 4
Gene Aliases:
6430526J12Rik, D230026E03, Megf7, mdig
Chromosome Location:
Chr.2: 91457470 - 91513901 on Build GRCm38
UniGene ID:
Mm.275149
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS213553
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrp4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172668.3 NP_766256.3 14 1944
GenBank mRNA AF247637.1 - 14 1774
AK051967.1 - 4 3268
BC132240.1 - 14 1867 40130517

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

kidney development
regulation of protein phosphorylation
hair follicle development
endocytosis
multicellular organism development
protein localization
dorsal/ventral pattern formation
proximal/distal pattern formation
Wnt signaling pathway
cell differentiation
negative regulation of Wnt signaling pathway
negative regulation of ossification
embryonic limb morphogenesis
odontogenesis of dentin-containing tooth
embryonic digit morphogenesis
receptor clustering
Wnt signaling pathway involved in dorsal/ventral axis specification
dendrite morphogenesis
anatomical structure development
negative regulation of axonogenesis
synapse organization
synaptic growth at neuromuscular junction
protein heterotetramerization
canonical Wnt signaling pathway
limb development
skeletal muscle acetylcholine-gated channel clustering
negative regulation of canonical Wnt signaling pathway
postsynaptic membrane assembly
presynaptic membrane assembly
positive regulation of presynaptic membrane organization
calcium ion binding
protein binding
Wnt-protein binding
receptor tyrosine kinase binding
apolipoprotein binding
protein homodimerization activity
Wnt-activated receptor activity
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0