This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1110051M20RIK SIRNA Assays ›
Gene Symbol:
1110051M20Rik
Gene Name:
RIKEN cDNA 1110051M20 gene
Gene Aliases:
AI586322
Chromosome Location:
Chr.2: 91273240 - 91444713 on Build GRCm38
UniGene ID:
Mm.386861
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS213549
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1110051M20Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175123.4 NP_780332.1 4, 5 383
GenBank mRNA AK018339.1 - 4, 5 417
AK045977.1 - 4, 5 384
AK164320.1 - 4, 5 381
BC032255.1 - 4, 5 403
BC049648.1 - 3, 4 318
BC085239.1 - 4, 5 382 6440075
BC089006.1 - 4, 5 363 6843427

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0