This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP185 SIRNA Assays ›
Gene Symbol:
Zfp185
Gene Name:
zinc finger protein 185
Gene Aliases:
P1, Znf185
Chromosome Location:
Chr.X: 72987253 - 73031543 on Build GRCm38
UniGene ID:
Mm.1161
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS213163
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp185
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109043.1 NP_001102513.1 18 1599
NM_009549.3 NP_033575.3 18 1602
GenBank mRNA AK014497.1 - 7 650
AK048188.1 - 18 1580
BC039772.1 - 15 1187 4191053
EF494744.1 - 18 1582
U46687.1 - 17 1428

Back To Top

More Information


Gene Ontology Categories:

Process(es)

zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0