This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZBTB14 SIRNA Assays ›
Gene Symbol:
Zbtb14
Gene Name:
zinc finger and BTB domain containing 14
Gene Aliases:
ZF5, Zfp161, b2b1982Clo
Chromosome Location:
Chr.17: 69383181 - 69390544 on Build GRCm38
UniGene ID:
Mm.29434
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS213149
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zbtb14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009547.2 NP_033573.1 3 1541
GenBank mRNA AK037725.1 - 3 1662
AK082051.1 - 3 1682
AK136762.1 - 1637
AK145517.1 - 3 1337
AK146201.1 - 3 1341
BC052017.1 - 3 1541 5697838
L15325.1 - 2 1272

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0