This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AFAP1L2 SIRNA Assays ›
Gene Symbol:
Afap1l2
Gene Name:
actin filament associated protein 1-like 2
Gene Aliases:
AU041783, C86904, mKIAA1914
Chromosome Location:
Chr.19: 56912354 - 57008590 on Build GRCm38
UniGene ID:
Mm.226284
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS213002
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Afap1l2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177796.1 NP_001171267.1 10, 11 1428
NM_001177797.1 NP_001171268.1 9, 10 1374
NM_146102.2 NP_666214.1 11, 12 1480
GenBank mRNA AK048649.1 - 11, 12 1134
AK087449.1 - 10, 11 1064
AK220430.1 - 9, 10 997
BC031515.1 - 10, 11 1030 3711746

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
regulation of mitotic cell cycle
regulation of signal transduction
regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of protein tyrosine kinase activity
SH3 domain binding
protein tyrosine kinase activator activity
signaling adaptor activity
SH2 domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0