This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIEZO2 SIRNA Assays ›
Gene Symbol:
Piezo2
Gene Name:
piezo-type mechanosensitive ion channel component 2
Gene Aliases:
5930434P17, 9030411M15Rik, 9430028L06Rik, Fam38b, Fam38b2
Chromosome Location:
Chr.18: 63010213 - 63387307 on Build GRCm38
UniGene ID:
Mm.158720
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS212843
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Piezo2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039485.4 NP_001034574.4 43, 44 7619
GenBank mRNA AK031235.1 - 3, 4 557
AK031422.1 - 2, 3 278
BC075677.1 - 4, 5 710
BC147606.1 - 2, 3 172 9056596
BC172109.1 - 2, 3 172
HQ215521.1 - 43, 44 6699

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
response to mechanical stimulus
regulation of membrane potential
response to stimulus
detection of mechanical stimulus involved in sensory perception
cation transmembrane transport
cation channel activity
protein binding
mechanically-gated ion channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0