This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MPPED1 SIRNA Assays ›
Gene Symbol:
Mpped1
Gene Name:
metallophosphoesterase domain containing 1
Gene Aliases:
6230403M19, AW456965, D15Bwg0669e
Chromosome Location:
Chr.15: 83779472 - 83858487 on Build GRCm38
UniGene ID:
Mm.36395
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS212444
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mpped1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172610.3 NP_766198.1 2 250
GenBank mRNA AK048421.1 - 2 251
AK158740.1 - 2 269
AK159045.1 - 2 367
AK165755.1 - 2 211
BC057550.1 - 1 175 6810744

Back To Top

More Information


Panther Classification:

Molecular Function -

esterase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
nuclease activity
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0