This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM184B SIRNA Assays ›
Gene Symbol:
Tmem184b
Gene Name:
transmembrane protein 184b
Gene Aliases:
2610507A11, 4732495E13Rik
Chromosome Location:
Chr.15: 79360684 - 79403303 on Build GRCm38
UniGene ID:
Mm.323358
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS212433
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem184b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253817.1 NP_001240746.1 9 1183
NM_001253819.1 NP_001240748.1 10 1225
NM_001253820.1 NP_001240749.1 7 749
NM_172608.1 NP_766196.1 9 1229
GenBank mRNA AK028196.1 - 9 1187
AK029129.1 - 9 1207
AK048138.1 - 10 1204
AK149366.1 - 9 1210
AK158789.1 - 9 1230
AK170086.1 - 9 1207
BC046959.1 - 9 1183 6314456

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0