This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNK9 SIRNA Assays ›
Gene Symbol:
Kcnk9
Gene Name:
potassium channel, subfamily K, member 9
Gene Aliases:
Task3
Chromosome Location:
Chr.15: 72502927 - 72547853 on Build GRCm38
UniGene ID:
Mm.386856
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS212399
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnk9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033876.1 NP_001029048.1 2 576
GenBank mRNA BC139167.1 - 2 637 8862189
BC139168.1 - 2 637 8862190
DQ185133.1 - 2 576

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
stabilization of membrane potential
regulation of ion transmembrane transport
cellular potassium ion transport
potassium ion transmembrane transport
voltage-gated ion channel activity
voltage-gated potassium channel activity
potassium channel activity
potassium ion leak channel activity
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header