This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TLL1 SIRNA Assays ›
Gene Symbol:
Tll1
Gene Name:
tolloid-like
Gene Aliases:
Tll, Tll-1
Chromosome Location:
Chr.8: 64014150 - 64205993 on Build GRCm38
UniGene ID:
Mm.418367
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS211899
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tll1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009390.2 NP_033416.2 19 3128
GenBank mRNA AK139103.1 - 7 917
U34042.1 - 19 3128

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
multicellular organism development
cell differentiation
metalloendopeptidase activity
calcium ion binding
collagen binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0