This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THEG SIRNA Assays ›
Gene Symbol:
Theg
Gene Name:
testicular haploid expressed gene
Gene Aliases:
-
Chromosome Location:
Chr.10: 79576477 - 79587136 on Build GRCm38
UniGene ID:
Mm.89197
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS211750
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Theg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011583.3 NP_035713.1 2 450
NM_199226.1 NP_954696.1 2 450
GenBank mRNA AB033128.1 - 2 432
AJ011834.1 - 2 635
BC050970.1 - 2 451
BC061243.1 - 2 396 6775002

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
cell differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0