This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TERT SIRNA Assays ›
Gene Symbol:
Tert
Gene Name:
telomerase reverse transcriptase
Gene Aliases:
EST2, TCS1, TP2, TR, TRT
Chromosome Location:
Chr.13: 73626797 - 73649843 on Build GRCm38
UniGene ID:
Mm.10109
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS211506
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tert
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009354.1 NP_033380.1 3 1668
GenBank mRNA AF051911.1 - 3 1668
AF073311.1 - 3 1639
BC082327.1 - 4 1170
BC127068.1 - 4 956 40110173

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, RNA-templated
RNA-dependent DNA biosynthetic process
telomere maintenance via telomerase
mitochondrion organization
negative regulation of gene expression
DNA strand elongation
positive regulation of Wnt signaling pathway
production of siRNA involved in RNA interference
RNA biosynthetic process
positive regulation of hair cycle
positive regulation of nitric-oxide synthase activity
cellular response to hypoxia
DNA biosynthetic process
replicative senescence
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
negative regulation of production of siRNA involved in RNA interference
positive regulation of stem cell proliferation
negative regulation of cellular senescence
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
tRNA binding
transcription coactivator binding
DNA binding
telomerase activity
telomerase RNA reverse transcriptase activity
RNA binding
RNA-directed DNA polymerase activity
RNA-directed RNA polymerase activity
protein binding
transferase activity
nucleotidyltransferase activity
telomeric DNA binding
protein homodimerization activity
metal ion binding
telomerase RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0