This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALX1 SIRNA Assays ›
Gene Symbol:
Alx1
Gene Name:
ALX homeobox 1
Gene Aliases:
AI314867, C130005I02, Cart1
Chromosome Location:
Chr.10: 103007847 - 103029452 on Build GRCm38
UniGene ID:
Mm.19287
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS211107
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Alx1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172553.4 NP_766141.1 1 277
GenBank mRNA AK047846.1 - 1 278
AK086731.1 - 1 90
BC052200.1 - 1 259 6513129

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006982, BC010923, U31986.
This siRNA also targets the following transcripts in Rat: NM_012921, XM_006241293, L14018, BC158591.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural tube closure
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
anterior/posterior pattern specification
positive regulation of epithelial to mesenchymal transition
mesenchymal cell development
embryonic limb morphogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
palate development
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0