This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBXAS1 SIRNA Assays ›
Gene Symbol:
Tbxas1
Gene Name:
thromboxane A synthase 1, platelet
Gene Aliases:
CYP5, CYP5A1, THAS, TS, TXAS, TXS
Chromosome Location:
Chr.6: 38918467 - 39084590 on Build GRCm38
UniGene ID:
Mm.4054
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS210635
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbxas1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011539.3 NP_035669.3 4 426
GenBank mRNA AK048544.1 - 4 408
AK139479.1 - 4 389
BC029014.1 - 4 428 5007629
L18868.1 - 4 297

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

prostaglandin biosynthetic process
lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
prostaglandin metabolic process
cellular chloride ion homeostasis
positive regulation of vasoconstriction
oxidation-reduction process
monooxygenase activity
thromboxane-A synthase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
isomerase activity
heme binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0