This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBR1 SIRNA Assays ›
Gene Symbol:
Tbr1
Gene Name:
T-box brain gene 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 61802734 - 61814114 on Build GRCm38
UniGene ID:
Mm.308525
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS210589
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009322.3 NP_033348.2 1 531
GenBank mRNA AK158835.1 - 1 450
BC052737.1 - 1 531 6825394
BC058399.1 - 1 435 6826831
U49251.1 - 1 371

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006593, BC029289, AK312567, BC113418, BC104844, and others.
This siRNA also targets the following transcripts in Rat: NM_001191070.

Gene Ontology Categories:

Function(s) Process(es)

conditioned taste aversion
transcription, DNA-templated
regulation of transcription, DNA-templated
specification of organ identity
regulation of gene expression
regulation of neuron projection development
amygdala development
commitment of neuronal cell to specific neuron type in forebrain
neuron differentiation
hindbrain development
positive regulation of transcription from RNA polymerase II promoter
regulation of axon guidance
RNA polymerase II core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0