This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EVI5L SIRNA Assays ›
Gene Symbol:
Evi5l
Gene Name:
ecotropic viral integration site 5 like
Gene Aliases:
1700084G18Rik, 2310039H16Rik, 3110007G05Rik, B130050I23Rik
Chromosome Location:
Chr.8: 4166538 - 4208050 on Build GRCm38
UniGene ID:
Mm.437106
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS210410
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Evi5l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039578.3 NP_001034667.3 4 640
GenBank mRNA AK045240.1 - 4 641
BC042653.1 - 4 620

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006248803, XM_008769048, FQ211110.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
GTPase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0