This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SORBS1 SIRNA Assays ›
Gene Symbol:
Sorbs1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
2310065E01Rik, 9530001P15Rik, CAP, SH3P12, Sh3d5, mKIAA1296
Chromosome Location:
Chr.19: 40292040 - 40513876 on Build GRCm38
UniGene ID:
Mm.210815
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS208975
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sorbs1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034962.1 NP_001030134.1 18, 19 1704
NM_001034963.1 NP_001030135.1 18, 19 1734
NM_001034964.1 NP_001030136.1 17, 18 1697
NM_009166.3 NP_033192.2 19, 20 1893
NM_178362.1 NP_848139.1 24, 25 2742
GenBank mRNA AF078666.1 - 18, 19 1723
AF078667.1 - 18, 19 1693
AF521593.1 - 16, 17 1654
AK035212.1 - 20, 21 2236
AK087553.1 - 24, 25 2744
AK122487.2 - 22, 23 3466
AK146712.1 - 16, 17 1532
AK207719.1 - 1, 2 83
BC012703.1 - 19, 20 1726 3602997
U58883.1 - 19, 20 1831

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008760454, XM_008760453, XM_008760456, XM_008760457, XM_008760458, and others.

Gene Ontology Categories:

Function(s) Process(es)

transport
actin filament organization
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
transcription factor binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0