This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SORBS1 SIRNA Assays ›
Gene Symbol:
Sorbs1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
2310065E01Rik, 9530001P15Rik, CAP, SH3P12, Sh3d5, mKIAA1296
Chromosome Location:
Chr.19: 40292040 - 40513876 on Build GRCm38
UniGene ID:
Mm.210815
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS208974
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sorbs1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034962.1 NP_001030134.1 22, 23 2203
NM_001034963.1 NP_001030135.1 22, 23 2233
NM_001034964.1 NP_001030136.1 21, 22 2196
NM_009166.3 NP_033192.2 23, 24 2392
NM_178362.1 NP_848139.1 28, 29 3241
GenBank mRNA AF078666.1 - 22, 23 2222
AF078667.1 - 22, 23 2192
AF521593.1 - 20, 21 2153
AK087553.1 - 28, 29 3243
AK122487.2 - 26, 27 3965
AK146712.1 - 20, 21 2030
BC012703.1 - 23, 24 2225 3602997
U58883.1 - 23, 24 2330

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
actin filament organization
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
transcription factor binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0