This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FOXP3 SIRNA Assays ›
Gene Symbol:
Foxp3
Gene Name:
forkhead box P3
Gene Aliases:
JM2, scurfin, sf
Chromosome Location:
Chr.X: 7579676 - 7595243 on Build GRCm38
UniGene ID:
Mm.182291
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS208941
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Foxp3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199347.1 NP_001186276.1 13 1414
NM_001199348.1 NP_001186277.1 12 1272
NM_054039.2 NP_473380.1 12 1347
GenBank mRNA AF277991.1 - 12 1272
AF277992.1 - 12 1330
AK157447.1 - 13 1415
AY357712.1 - 11 1130
AY357713.1 - 11 1130
BC132333.1 - 11 1118 40130610
BC132335.1 - 11 1118 40130612
DQ387959.1 - 13 1414
KF471319.1 - 11 1114

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
B cell homeostasis
cytokine production
myeloid cell homeostasis
CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
CD4-positive, CD25-positive, alpha-beta regulatory T cell lineage commitment
T cell mediated immunity
tolerance induction
tolerance induction to self antigen
regulation of immunoglobulin production
positive regulation of T cell tolerance induction
regulation of T cell anergy
positive regulation of T cell anergy
negative regulation of chronic inflammatory response
negative regulation of T cell cytokine production
positive regulation of peripheral T cell tolerance induction
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
negative regulation of cell proliferation
anatomical structure morphogenesis
positive regulation of gene expression
negative regulation of histone deacetylation
negative regulation of NF-kappaB transcription factor activity
negative regulation of interferon-gamma production
negative regulation of interleukin-10 production
negative regulation of interleukin-17 production
negative regulation of interleukin-2 production
negative regulation of interleukin-4 production
negative regulation of interleukin-5 production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
positive regulation of interleukin-4 production
negative regulation of CREB transcription factor activity
positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
positive regulation of transforming growth factor beta1 production
positive regulation of immature T cell proliferation in thymus
positive regulation of histone acetylation
negative regulation of histone acetylation
negative regulation of cytokine biosynthetic process
T cell activation
negative regulation of T cell proliferation
negative regulation of sequence-specific DNA binding transcription factor activity
regulatory T cell differentiation
negative regulation of interferon-gamma biosynthetic process
negative regulation of interleukin-2 biosynthetic process
positive regulation of regulatory T cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of isotype switching to IgE isotypes
regulation of isotype switching to IgG isotypes
negative regulation of lymphocyte proliferation
negative regulation of cytokine secretion
negative regulation of inflammatory response
negative regulation of immune response
T cell receptor signaling pathway
negative regulation of T-helper 17 cell differentiation
core promoter binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
histone acetyltransferase binding
protein homodimerization activity
histone deacetylase binding
sequence-specific DNA binding
metal ion binding
NFAT protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0